Learn More
Medchemexpress LLC Trimethyl-Histone H3 (Lys27) antibody | 50 UL
SDP
Catalog No. 5000443000
Encompass_Preferred
TriMethyl-Histone H3 (Lys27) Antibody is a recombinant, non-conjugated IgG monoclonal antibody derived from Rabbit, specifically targeting TriMethyl-Histone H3 (Lys27). This antibody is for research use only and is supplied as a liquid solution containing 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.05% BSA, and 0.01% Sodium azide as a preservative.
  • Endogenous sensitivity
  • Affinity purified
  • Reacts with human and rat species
  • Immunogen is a synthetic peptide corresponding to residues surrounding Lys27 of Human Histone H3
  • Applicable in Western Blot, Immunohistochemistry (Paraffin and Frozen), Immunocytochemistry/Immunofluorescence, Immunoprecipitation, and Chromatin Immunoprecipitation
Catalog No. 50-004-43000 Supplier Medchemexpress LLC Supplier No. HYP8092250UL
TriMethyl-Histone H3 (Lys27) Antibody is a recombinant, non-conjugated IgG monoclonal antibody derived from Rabbit, specifically targeting TriMethyl-Histone H3 (Lys27). This antibody is for research use only and is supplied as a liquid solution containing 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.05% BSA, and 0.01% Sodium azide as a preservative.
  • Endogenous sensitivity
  • Affinity purified
  • Reacts with human and rat species
  • Immunogen is a synthetic peptide corresponding to residues surrounding Lys27 of Human Histone H3
  • Applicable in Western Blot, Immunohistochemistry (Paraffin and Frozen), Immunocytochemistry/Immunofluorescence, Immunoprecipitation, and Chromatin Immunoprecipitation