Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl(nonafluorobutyl)silane 98.0+%, TCI America™
SDP

Catalog No. T35945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35945G 5 g
T35941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35945G Supplier TCI America Supplier No. T35945G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 204316-01-8
Molecular Formula C7H9F9Si
Molecular Weight (g/mol) 292.22
MDL Number MFCD20489405
InChI Key YHSOLWOJHMOEFX-UHFFFAOYSA-N
Synonym Trimethyl(perfluorobutyl)silane
PubChem CID 15534208
IUPAC Name trimethyl(1,1,2,2,3,3,4,4,4-nonafluorobutyl)silane
SMILES C[Si](C)(C)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 100°C
Quantity 5 g
UN Number 1993
Formula Weight 292.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl(nonafluorobutyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.