Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyl Orthoformate

Catalog No. 3054722L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-547-22L 2 L
30-547-2100ML 100 mL
30-547-21L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 30-547-22L Supplier Sigma Aldrich Supplier No. 3054722L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethyl orthoformate is a commonly used reagent in organic synthesis for the preparation of useful building blocks. It is also used to introduce the protecting group for aldehydes by the preparation of acetals. Acetals can be deprotected back to the aldehyde by using acid catalysts.Trimethyl orthoformate can be used:To convert sulfonic acids to methyl esters.To convert 2-acylcyclohexanones to the corresponding acetal derivatives.To mediate Pinacol reaction of various 1,2-diols with tin(IV) chloride without the formation of water.To synthesize 1-substituted-1H-1,2,3,4-tetrazoles via a three-component condensation with amine and sodium azide catalyzed by indium triflate under solvent-free conditions.For the N-methylation of amines in the presence of sulfuric acid.

Specifications

CAS 149-73-5
Density 0.97 g/mL at 25°C
Boiling Point 101°C to 102°C
Molecular Formula C4H10O3
Refractive Index n20/D 1.379
Linear Formula CH(OCH3)3
MDL Number MFCD00008483
Quantity 2 L
Synonym TMOF; Trimethoxymethane
Molecular Weight (g/mol) 106.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.