Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl(pentamethylcyclopentadienyl)titanium(IV), 97%

Catalog No. AA4375902
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4375902 0.5 g
AA4375904 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4375902 Supplier Thermo Scientific Chemicals Supplier No. 04375902
Add to Cart

CAS: 107333-47-1 | C13H29Ti-5 | 233.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107333-47-1
Molecular Formula C13H29Ti-5
Molecular Weight (g/mol) 233.242
MDL Number MFCD00269851
InChI Key DVHMCUKDWMNRGX-UHFFFAOYSA-N
Synonym acmc-1bo0g, titanium,trimethyl 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
PubChem CID 57346580
IUPAC Name carbanide;cyclopenta-1,3-diene;titanium(4+)
SMILES [CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Ti+4]

Specifications

Packaging Ampule
Quantity 0.5 g
UN Number UN1325
Sensitivity Air, light, and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 228.22
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Trimethyl (pentamethylcyclopentadienyl)titanium (IV)
Hazard Category H228-H319-H335
Hazard Statement GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.