Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimethyl phosphate | 512-56-1 | 98.0% | C3H9O4P | 1 ML
SDP

Supplier:  Medchemexpress LLC HYA0108A10MM/1ML

Encompass_Preferred

Trimethyl phosphate is a biochemical reagent that can be utilized as a biological material or an organic compound for life science related research.

  • Suitable as a biological material or organic compound
  • Utilized for life science related research
  • Purity of 98.0%
  • Molecular weight of 140.07
  • Appears as a colorless to light yellow liquid
  • Available as a 10 mM solution in 1 mL of DMSO
  • Store at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-32060


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.