Learn More
Medchemexpress LLC Trimethyl phosphate-d9 | 32176-12-8 | 99.4% | C3D9O4P | 100 MG
SDP
Catalog No. 5000432219
Encompass_Preferred
Trimethyl phosphate-d9 is the deuterium labeled version of Trimethyl phosphate (HY-A0108A). Trimethyl phosphate acts as a methylating agent, transferring methyl groups to aliphatic alcohols, phenols, and amines. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32219 Supplier Medchemexpress LLC Supplier No. HYA0108S100MG
Trimethyl phosphate-d9 is the deuterium labeled version of Trimethyl phosphate (HY-A0108A). Trimethyl phosphate acts as a methylating agent, transferring methyl groups to aliphatic alcohols, phenols, and amines. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS