Learn More
Medchemexpress LLC Trimethyl phosphate-d9 | 32176-12-8 | 99.4% | C3D9O4P | 50 MG
SDP
Catalog No. 5000432696
Encompass_Preferred
Trimethyl phosphate-d9 is the deuterium labeled version of Trimethyl phosphate. It acts as a methylating agent, transferring methyl groups to aliphatic alcohols, phenols, and amines. This compound is useful as a tracer or as an internal standard in quantitative analysis techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, known as deuteration, has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-32696 Supplier Medchemexpress LLC Supplier No. HYA0108S50MG
Trimethyl phosphate-d9 is the deuterium labeled version of Trimethyl phosphate. It acts as a methylating agent, transferring methyl groups to aliphatic alcohols, phenols, and amines. This compound is useful as a tracer or as an internal standard in quantitative analysis techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, known as deuteration, has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs