Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyl Phosphonoacetate

Catalog No. T79758100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T79758100G 100 g
T7975825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T79758100G Supplier Sigma Aldrich Supplier No. T79758100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in:Intramolecular Mannich-type reactions to produce the sarain A diazatricyclic coreOrganocatalytic oxa-Michael reactionsPrenylation and geranylation of oxindolesIntramolecular Horner-Wadsworth-Emmons reactionsOlefin cross-metathesis / heterocyclization

Specifications

CAS 5927-18-4
Density 1.125 g/mL at 25°C
Boiling Point 118°C (0.85 mmHg, lit.), 265°C to 268°C
Molecular Formula C5H11O5P
Refractive Index n20/D 1.437
Linear Formula (CH3O)2P(O)CH2CO2CH3
MDL Number MFCD00008452
Quantity 100 g
Synonym (Carboxymethyl)phosphonic acid trimethyl ester; NSC 84262
Molecular Weight (g/mol) 182.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.