Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl thiophosphate

Catalog No. p-7049862
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3039503 1 g
AA3039506 5 g
AA3039514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3039503 Supplier Thermo Scientific Chemicals Supplier No. 03039503
Only null left
Add to Cart

CAS: 152-18-1 | C3H9O3PS | 156.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-18-1
Molecular Formula C3H9O3PS
Molecular Weight (g/mol) 156.136
MDL Number MFCD00014888
InChI Key XWSLYQXUTWUIKM-UHFFFAOYSA-N
Synonym trimethyl thiophosphate, trimethylthiophosphate, o,o,o-trimethyl phosphorothioate, o,o,o-trimethyl thiophosphate, trimethylthiofosfat, o,o,o-trimethylthiofosfat, trimethylthiofosfat czech, ccris 5569, phosphorothioic acid, o,o,o-trimethyl ester, o,o,o-trimethylthiofosfat czech
PubChem CID 9038
IUPAC Name trimethoxy(sulfanylidene)-$l^{5}-phosphane
SMILES COP(=S)(OC)OC

Specifications

Color White
Odor Sulphurous
Quantity 1 g
UN Number UN3278
Formula Weight 156.15
Physical Form Liquid
Chemical Name or Material Trimethyl thiophosphate
Hazard Category H302-H315-H319-H330-H335
Hazard Statement GHS H Statement
H330-H302-H315-H319-H335
Fatal if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.