Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyl(Trifluoromethyl)Silane Solution

Catalog No. 9186210ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-862-10ML 10 mL
91-862-50ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 91-862-10ML Supplier Sigma Aldrich Supplier No. 9186210ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethyl(trifluoromethyl)silane (TMSCF3) is also called as Ruppert-Prakash fluorination reagent. It is extensively used for the synthesis of trifluoromethyl-containing compounds.TMSCF3 can be used as a reagent for:Conversion of aromatic aldehydes to difluoromethylated products.C-H trifluoromethylation of arenes, terminal alkynes, tertiary amines, heteroarenes, allylic, and terminal alkenes using metal catalyst or metal free oxidative trifluoromethylation reaction.It can also be used in trifluoromethylation of: Non-activated aldimines.Heterocumulenes.Azomethine imines.

Specifications

CAS 81290-20-2
Density 0.91 g/mL at 20°C
Molecular Formula C4H9F3Si
Linear Formula (CH3)3SiCF3
Refractive Index n20/D 1.386
Quantity 10 mL
MDL Number MFCD00145454
Synonym (Trifluoromethyl)trimethylsilane; Ruppert's reagent; Ruppert-Prakash reagent; TFMTMS; Trifluoromethyltrimethylsilane
Molecular Weight (g/mol) 142.19 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.