Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl Trimellitate 95.0+%, TCI America™
SDP

Catalog No. T122925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T122925G 25 g
T1229500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T122925G Supplier TCI America Supplier No. T122925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10-1-2459
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.22
MDL Number MFCD00043629
InChI Key MJHNUUNSCNRGJE-UHFFFAOYSA-N
Synonym trimethyl trimellitate, trimethyl 1,2,4-benzenetricarboxylate, methyl trimellitate, trimellitic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, trimethyl ester, unii-16o7dt5n66, 1,2,4-benzenetricarboxylic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester, 1,2,4-trimethyl benzene-1,2,4-tricarboxylate, acmc-1ckkm
PubChem CID 17159
IUPAC Name trimethyl benzene-1,2,4-tricarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)C(=O)OC

Specifications

Color Yellow
Boiling Point 189°C
Quantity 25 g
Formula Weight 252.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Trimellitate
EINECSNumber (3)-1371
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.