Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylacetaldehyde

Catalog No. T715015ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T715015ML 5 mL
T71501100ML 100 mL
T7150125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T715015ML Supplier Sigma Aldrich Supplier No. T715015ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylacetaldehyde

Specifications

CAS 630-19-3
Melting Point 6°C (lit.)
Density 0.793 g/mL (at 25°C (literature))
Boiling Point 74°C (730 mmHg)
Molecular Formula C5H10O
Refractive Index n20/D 1.378 (literature)
Linear Formula (CH3)3 CCHO
MDL Number MFCD00006962
Quantity 5 mL
Synonym Pivalaldehyde; Trimethylacetaldehyde
Molecular Weight (g/mol) 86.13
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.