Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylacetic acid, 99%

Catalog No. p-206894
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140020050 5 g Glass bottle
AC140021000 100 g Glass bottle
AC140025000 500 g Glass bottle
AC140020025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC140020050 Supplier Thermo Scientific Chemicals Supplier No. 140020050
Only null left
Add to Cart

CAS: 75-98-9 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-98-9
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00004194
InChI Key IUGYQRQAERSCNH-UHFFFAOYSA-N
Synonym pivalic acid, trimethylacetic acid, neopentanoic acid, 2,2-dimethylpropionic acid, tert-pentanoic acid, propanoic acid, 2,2-dimethyl, versatic 5, kyselina pivalova, acetic acid, trimethyl, alpha,alpha-dimethylpropionic acid
PubChem CID 6417
ChEBI CHEBI:45133
IUPAC Name 2,2-dimethylpropanoic acid
SMILES CC(C)(C)C(=O)O

Specifications

Melting Point 33.0°C to 35.0°C
Color Tan to Yellow
Boiling Point 163.0°C to 164.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCO2H
Quantity 5 g
Beilstein 02, 319
Fieser 01,1229
Merck Index 15, 7625
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 3.6 mm2/s (60°C)
Formula Weight 102.13
Percent Purity 99%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Trimethylacetic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
EINECSNumber 200-922-5
RTECSNumber TO7700000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.