Learn More
Medchemexpress LLC Trimethylacetohydrazideammonium chloride | 123-46-6 | MFCD00012009 | 99.8% | 167.64 g/mol | C5H14ClN3O | 1 KG
SDP
Catalog No. 5000210814
Encompass_Preferred
Girard's reagent T is a derivatizing agent that reacts with aldehydes and ketones to form stable hydrazones, facilitating analysis by chromatography and spectrophotometry and used in analytical and synthetic applications.
  • Reacts selectively with carbonyl compounds to form stable, analyzable hydrazones.
  • Facilitates chromatographic and spectrophotometric detection of aldehydes and ketones.
  • High purity suitable for analytical and synthetic use.
  • Available in multiple laboratory pack sizes for scalable applications.
Catalog No. 50-002-10814 Supplier Medchemexpress LLC Supplier No. HYW0745411KG
Girard's reagent T is a derivatizing agent that reacts with aldehydes and ketones to form stable hydrazones, facilitating analysis by chromatography and spectrophotometry and used in analytical and synthetic applications.
  • Reacts selectively with carbonyl compounds to form stable, analyzable hydrazones.
  • Facilitates chromatographic and spectrophotometric detection of aldehydes and ketones.
  • High purity suitable for analytical and synthetic use.
  • Available in multiple laboratory pack sizes for scalable applications.