Learn More
Medchemexpress LLC Trimethylacetohydrazideammonium chloride | 123-46-6 | MFCD00012009 | 99.8% | 167.64 g/mol | C5H14ClN3O | 100 G
SDP
Supplier:  Medchemexpress LLC HYW074541100G
Encompass_Preferred
Girard's reagent T is a quaternary hydrazide salt used to derivatize carbonyl compounds (aldehydes and ketones) into stable hydrazones for chromatographic and spectrometric analysis. It is a water-soluble solid commonly used in analytical chemistry and organic synthesis for selective enrichment, detection, and quantification of carbonyl-containing molecules.
  • Derivatizes aldehydes and ketones to form stable hydrazones.
  • Water-soluble solid suitable for chromatographic and spectrometric analysis.
  • High purity for reliable analytical results.
  • Useful for selective enrichment and detection of carbonyl-containing compounds.
  • Available in multiple pack sizes for laboratory and bulk use.
Catalog No. 50-002-09796