Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylacetyl chloride

Catalog No. T72605100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T72605100ML 100 mL
T72605500ML 500 mL
T726055ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T72605100ML Supplier Sigma Aldrich Supplier No. T72605100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Widely used N-acylating agent for amines, Schiff bases, and pyrrolidinones as well as O-acylating agent for alcohols, lactones, and saccharides.

Specifications

CAS 3282-30-2
Density 0.979 g/mL (at 25°C (literature))
Boiling Point 105°C to 106°C (lit.)
Molecular Formula C5H9ClO
Refractive Index n20/D 1.412 (literature)
Linear Formula (CH3)3 CCOCl
MDL Number MFCD00000709
Quantity 100 mL
Synonym Pivaloyl chloride; Trimethylacetyl chloride
Molecular Weight (g/mol) 120.58
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.