Learn More
Strem, An Ascensus Company CAS# 75-24-1. 25g. Trimethylaluminum, min. 98%, 93-1360, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00008252
SDP
Supplier:  Strem, An Ascensus Company 98400325G
Encompass
CAS# 75-24-1. 25g. Trimethylaluminum, min. 98%, 93-1360, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00008252. Molecular Weight: 72.09. Molecular Formula: (CH3)3Al. Color/form: colorless liq. Strem# 98-4003. http://www.strem.com/catalog/v/98-4003/
Catalog No. 50-201-6760