Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylaluminium, 1.0M solution in heptane, AcroSeal™

Catalog No. p-2799239
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189271000 100 mL AcroSeal™ Glass bottle
AC189278000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC189271000 Supplier Thermo Scientific Chemicals Supplier No. 189271000
Only null left
Add to Cart

CAS: 75-24-1 | C3H9Al | 72.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-24-1 , 142-82-5
Molecular Formula C3H9Al
Molecular Weight (g/mol) 72.087
MDL Number MFCD00008252
InChI Key JLTRXTDYQLMHGR-UHFFFAOYSA-N
Synonym trimethylaluminum, trimethylaluminium, aluminum, trimethyl, trimethylalane, unii-av210lg46j, ch3 3al, aluminum trimethanide, aluminum trimethyl, trimethylaluminum solution, trimethylaluminum, elec. gr.
PubChem CID 16682925
IUPAC Name trimethylalumane
SMILES C[Al](C)C

Specifications

Color Colorless
Boiling Point 126.0°C
Melting Point 15.0°C
Linear Formula (CH3)3Al
Fieser 08,506; 14,46; 17,372
Merck Index 15, 9879
Density 0.6880g/mL
Solubility Information Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
Formula Weight 72.09
Percent Purity 9.5 to 10.5 wt% (on Al basis)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.688
Viscosity 0.90 mPa.s (30°C)
Quantity 100 mL
Flash Point −4°C
Physical Form Liquid
Chemical Name or Material Trimethylaluminium, 1.0M Solution in Heptane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.