Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylaluminum

Catalog No. 257222100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-722-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-722-2100G Supplier Sigma Aldrich Supplier No. 257222100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylaluminum

Specifications

CAS 75-24-1
Melting Point 15°C
Density 0.752 g/mL (at 25°C)
Boiling Point 125°C to 126°C; 127°C (760 mmHg); 20°C (8 mmHg); 56°C (50 mmHg)
Molecular Formula C3H9Al
Linear Formula (CH3)3 Al
Quantity 100 g
MDL Number MFCD00008252
Synonym Aluminum trimethanide; TMA
Molecular Weight (g/mol) 72.09
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.