Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylaluminum Solution

Catalog No. 379239800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-923-9800ML 800 mL
37-923-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 37-923-9800ML Supplier Sigma Aldrich Supplier No. 379239800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylaluminum can be used in the synthesis of methylaluminoxane (MAO), a precatalyst for olefin polymerization. It is a precursor for the preparation of a wide range of organoaluminium such as dimethylaluminum chloride and bis(trimethylaluminum)-1,4-diazabicyclo[2.2.2]-octane (DABAL-Me3).

Specifications

CAS 75-24-1
Density 0.693 g/mL at 25°C
Molecular Formula C3H9Al
Linear Formula (CH3)3Al
MDL Number MFCD00008252
Quantity 800 mL
Synonym Aluminum trimethanide solution
Molecular Weight (g/mol) 72.09 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.