Learn More
Cayman Chemical TrImethylamIn N-oxIde 1g
SDP
Supplier:  Cayman Chemical 173541 G
Encompass_Preferred
A metabolite of choline, phosphatidylcholine, and L-carnitine; dietary administration increases renal tubulointerstitial fibrosis, collagen deposition, and Smad3 phosphorylation in mice and increases aortic lesion area in atherosclerosis-prone ApoE-/- mice at 0.12% w/w; plasma levels of TMAO are elevated in patients with chronic kidney disease and decreased in patients with active, compared with inactive, ulcerative colitis; elevated plasma levels of TMAO are associated with increased risk of cardiovascular disease
Catalog No. 50-296-5205