Learn More
Medchemexpress LLC Trimethylamine hydrochloride | 593-81-7 | MFCD00012478 | 95.57 | C3H10ClN | 10g
SDP
Catalog No. 5000268431
Encompass_Preferred
Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]
Catalog No. 50-002-68431 Supplier Medchemexpress LLC Supplier No. HYY050410G
Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]