Learn More
Medchemexpress LLC Trimethylamine hydrochloride | 593-81-7 | MFCD00012478 | 95.57 g/mol | C3H10ClN | 250mg
SDP
Catalog No. 5000269175
Encompass_Preferred
Trimethylammonium chloride (Standard) is the analytical standard of Trimethylammonium chloride This product is intended for research and analytical applications Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]
Catalog No. 50-002-69175 Supplier Medchemexpress LLC Supplier No. HYY0504R250MG
Trimethylammonium chloride (Standard) is the analytical standard of Trimethylammonium chloride This product is intended for research and analytical applications Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]