Learn More
Medchemexpress LLC Trimethylamine hydrochloride | 593-81-7 | MFCD00012478 | 95.57 g/mol | C3H10ClN | 50g
SDP
Catalog No. 5000268887
Encompass_Preferred
Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]
Catalog No. 50-002-68887 Supplier Medchemexpress LLC Supplier No. HYY050450G
Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]