Learn More
Medchemexpress LLC Trimethylamine hydrochloride | 593-81-7 | MFCD00012478 | 98.0% | 95.57 g/mol | C3H10ClN | 1mL
SDP
Catalog No. 5000269320
Encompass_Preferred
Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]
Catalog No. 50-002-69320 Supplier Medchemexpress LLC Supplier No. HYY050410MM/1ML
Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]