Learn More
Medchemexpress LLC Trimethylamine N-oxi 10mg | 1161070-49-0 | 10 MG
SDP
Catalog No. 5000229889
Encompass_Preferred
Trimethylamine N-oxide-d9 is the deuterium-labeled internal standard of trimethylamine N-oxide (TMAO) used for quantitative analytical assays such as LC-MS and GC-MS. Supplied as a solid high-purity reference standard, it is intended for calibration, method validation, and trace-level quantification in biological and environmental samples.
  • Deuterium-labeled internal standard for accurate quantification by LC-MS and GC-MS.
  • High purity (99.76%) for reliable calibration and trace-level detection.
  • Available in small pack sizes suitable for analytical workflows (1 mg-100 mg).
  • Solid powder with recommended storage at -20°C for long-term stability; in solvent store at -80°C for best stability.
  • Molecular weight 84.17 and molecular formula C3D9NO for unambiguous identification.
Catalog No. 50-002-29889 Supplier Medchemexpress LLC Supplier No. HY116084S10MG
Trimethylamine N-oxide-d9 is the deuterium-labeled internal standard of trimethylamine N-oxide (TMAO) used for quantitative analytical assays such as LC-MS and GC-MS. Supplied as a solid high-purity reference standard, it is intended for calibration, method validation, and trace-level quantification in biological and environmental samples.
  • Deuterium-labeled internal standard for accurate quantification by LC-MS and GC-MS.
  • High purity (99.76%) for reliable calibration and trace-level detection.
  • Available in small pack sizes suitable for analytical workflows (1 mg-100 mg).
  • Solid powder with recommended storage at -20°C for long-term stability; in solvent store at -80°C for best stability.
  • Molecular weight 84.17 and molecular formula C3D9NO for unambiguous identification.