Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimethylamine N-oxi 10mM 1mL | 1184-78-7 | 75.11 g/mol | C3H9NO | 1 ML
SDP

Catalog No. 5000229485
Encompass_Preferred

Trimethylamine N-oxide (TMAO) is a small-molecule metabolite used in biochemical and cell biology research, including studies of inflammation and metabolic pathways. The product is supplied as a 10 mM solution in a 1 mL pack for research applications.

  • Ready-to-use 10 mM solution, 1 mL.
  • Purity: 98.0%.
  • Molecular formula C3H9NO; molecular weight 75.11 g/mol.
  • Commonly used to study ROS and NLRP3 inflammasome activation.
  • COA and SDS available for quality and safety information.

Catalog No. 50-002-29485 Supplier Medchemexpress LLC Supplier No. HY11608410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Trimethylamine N-oxide (TMAO) is a small-molecule metabolite used in biochemical and cell biology research, including studies of inflammation and metabolic pathways. The product is supplied as a 10 mM solution in a 1 mL pack for research applications.

  • Ready-to-use 10 mM solution, 1 mL.
  • Purity: 98.0%.
  • Molecular formula C3H9NO; molecular weight 75.11 g/mol.
  • Commonly used to study ROS and NLRP3 inflammasome activation.
  • COA and SDS available for quality and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.