Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimethylamine N-oxi 1g | 1184-78-7 | 75.11 g/mol | C3H9NO | 1 G
SDP

Catalog No. 5000229282
Encompass_Preferred

Trimethylamine N-oxide (TMAO) is a small, water-soluble metabolite formed from dietary trimethylamine-containing nutrients and produced via gut microbial metabolism. Supplied as a purified solid, it is used in research on inflammation, ROS/NLRP3 inflammasome activation, and tissue fibrosis, and as a biochemical reagent in cell and molecular studies.

  • Purity 98.0% suitable for biochemical and cellular assays.
  • Molecular weight 75.11 g/mol; formula C3H9NO.
  • Available in common lab pack sizes, including 1 G solid packs.
  • Used to study inflammasome activation, fibrosis, and microbiome-host interactions.

Catalog No. 50-002-29282 Supplier Medchemexpress LLC Supplier No. HY1160841G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trimethylamine N-oxide (TMAO) is a small, water-soluble metabolite formed from dietary trimethylamine-containing nutrients and produced via gut microbial metabolism. Supplied as a purified solid, it is used in research on inflammation, ROS/NLRP3 inflammasome activation, and tissue fibrosis, and as a biochemical reagent in cell and molecular studies.

  • Purity 98.0% suitable for biochemical and cellular assays.
  • Molecular weight 75.11 g/mol; formula C3H9NO.
  • Available in common lab pack sizes, including 1 G solid packs.
  • Used to study inflammasome activation, fibrosis, and microbiome-host interactions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.