Learn More
Medchemexpress LLC Trimethylamine N-oxi 1mg | 1161070-49-0 | 84.17 | C3D9NO | 1 MG
SDP
Catalog No. 5000226420
Encompass_Preferred
Trimethylamine N-oxide-d9 is the deuterium-labeled analogue of trimethylamine N-oxide used as a stable isotope internal standard for quantitative mass spectrometry and metabolic studies. It provides a chemically matched, isotopically distinct reference for accurate LC-MS or GC-MS quantification of TMAO in biological samples.
  • Deuterated internal standard (d9) for trimethylamine N-oxide quantitation.
  • High isotopic purity, 99.76%.
  • Isotopic formula C3D9NO and molecular weight 84.17 g·mol⁻¹.
  • Available in small pack sizes suitable for analytical standards (1 mg-100 mg).
  • Solid form with recommended storage as powder at -20°C for long-term stability.
Catalog No. 50-002-26420 Supplier Medchemexpress LLC Supplier No. HY116084S1MG
Trimethylamine N-oxide-d9 is the deuterium-labeled analogue of trimethylamine N-oxide used as a stable isotope internal standard for quantitative mass spectrometry and metabolic studies. It provides a chemically matched, isotopically distinct reference for accurate LC-MS or GC-MS quantification of TMAO in biological samples.
  • Deuterated internal standard (d9) for trimethylamine N-oxide quantitation.
  • High isotopic purity, 99.76%.
  • Isotopic formula C3D9NO and molecular weight 84.17 g·mol⁻¹.
  • Available in small pack sizes suitable for analytical standards (1 mg-100 mg).
  • Solid form with recommended storage as powder at -20°C for long-term stability.