Learn More
Medchemexpress LLC Trimethylamine N-oxide dihydrate | 62637-93-8 | ≥98.0% | 111.14 | 25 G
SDP
Catalog No. 5000347583
Encompass_Preferred
Trimethylamine N-oxide dihydrate is a gut microbe-dependent metabolite derived from dietary choline and other trimethylamine-containing nutrients. It induces inflammation by activating the ROS/NLRP3 inflammasome. Additionally, it accelerates fibroblast-myofibroblast differentiation and triggers cardiac fibrosis through the activation of the TGF-β/smad2 signaling pathway. Trimethylamine N-oxide is also found in various marine organisms, where it offers protection against the adverse effects of temperature, salinity, high urea, and hydrostatic pressure.
  • Induces inflammation by activating the ROS/NLRP3 inflammasome.
  • Accelerates fibroblast-myofibroblast differentiation.
  • Triggers cardiac fibrosis through activation of the TGF-β/smad2 signaling pathway.
  • Provides protection against adverse effects of temperature, salinity, high urea, and hydrostatic pressure in marine organisms.
Catalog No. 50-003-47583 Supplier Medchemexpress LLC Supplier No. 25GHY108915
Trimethylamine N-oxide dihydrate is a gut microbe-dependent metabolite derived from dietary choline and other trimethylamine-containing nutrients. It induces inflammation by activating the ROS/NLRP3 inflammasome. Additionally, it accelerates fibroblast-myofibroblast differentiation and triggers cardiac fibrosis through the activation of the TGF-β/smad2 signaling pathway. Trimethylamine N-oxide is also found in various marine organisms, where it offers protection against the adverse effects of temperature, salinity, high urea, and hydrostatic pressure.
  • Induces inflammation by activating the ROS/NLRP3 inflammasome.
  • Accelerates fibroblast-myofibroblast differentiation.
  • Triggers cardiac fibrosis through activation of the TGF-β/smad2 signaling pathway.
  • Provides protection against adverse effects of temperature, salinity, high urea, and hydrostatic pressure in marine organisms.