Learn More
Medchemexpress LLC Trimethylamine N-oxide dihydrate | 62637-93-8 | 99.9% | 111.14 | 1 MG
SDP
Catalog No. 5000345841
Encompass_Preferred
Trimethylamine N-oxide dihydrate is a gut microbe-dependent metabolite implicated in various biological processes. It is known to induce inflammation via ROS/NLRP3 inflammasome activation and accelerate fibroblast-myofibroblast differentiation, contributing to cardiac fibrosis through the TGF-β/smad2 signaling pathway. This product is for research use only.
  • Induces inflammation and cardiac fibrosis.
  • Accelerates fibroblast-myofibroblast differentiation.
  • Modulates key signaling pathways including ROS/NLRP3 and TGF-β/smad2.
  • Used in animal modeling for cardiovascular and metabolic diseases.
  • Offers protective effects in marine organisms against environmental stressors.
Catalog No. 50-003-45841 Supplier Medchemexpress LLC Supplier No. HY10891510G
Trimethylamine N-oxide dihydrate is a gut microbe-dependent metabolite implicated in various biological processes. It is known to induce inflammation via ROS/NLRP3 inflammasome activation and accelerate fibroblast-myofibroblast differentiation, contributing to cardiac fibrosis through the TGF-β/smad2 signaling pathway. This product is for research use only.
  • Induces inflammation and cardiac fibrosis.
  • Accelerates fibroblast-myofibroblast differentiation.
  • Modulates key signaling pathways including ROS/NLRP3 and TGF-β/smad2.
  • Used in animal modeling for cardiovascular and metabolic diseases.
  • Offers protective effects in marine organisms against environmental stressors.