Learn More
Medchemexpress LLC Trimethylamine oxide-15N | 99.9% | 76.10 | C3H9^15NO | 10mg
SDP
Catalog No. 5000269437
Encompass_Preferred
Trimethylamine oxide-15N is the deuterium labeled Trimethylamine N-oxide (HY-116084) Trimethylamine N-oxide is a gut microbe-dependent metabolite of dietary choline and other trimethylamine-containing nutrients Trimethylamine N-oxide induces inflammation by activating the ROS/NLRP3 inflammasome Trimethylamine N-oxide also accelerates fibroblast-myofibroblast differentiation and induces cardiac fibrosis by activating the TGF- /smad2 signaling pathway[1][2][3]
Catalog No. 50-002-69437 Supplier Medchemexpress LLC Supplier No. HYW72256210MG
Trimethylamine oxide-15N is the deuterium labeled Trimethylamine N-oxide (HY-116084) Trimethylamine N-oxide is a gut microbe-dependent metabolite of dietary choline and other trimethylamine-containing nutrients Trimethylamine N-oxide induces inflammation by activating the ROS/NLRP3 inflammasome Trimethylamine N-oxide also accelerates fibroblast-myofibroblast differentiation and induces cardiac fibrosis by activating the TGF- /smad2 signaling pathway[1][2][3]