Learn More
Medchemexpress LLC Trimethylamine oxide-15N | MFCD00002048 | 99.9% | 76.10 | C3H9(15)NO | 1mg
SDP
Catalog No. 5000270493
Encompass_Preferred
Trimethylamine oxide-15N is the deuterium labeled Trimethylamine N-oxide (HY-116084) Trimethylamine N-oxide is a gut microbe-dependent metabolite of dietary choline and other trimethylamine-containing nutrients Trimethylamine N-oxide induces inflammation by activating the ROS/NLRP3 inflammasome Trimethylamine N-oxide also accelerates fibroblast-myofibroblast differentiation and induces cardiac fibrosis by activating the TGF- /smad2 signaling pathway[1][2][3]
Catalog No. 50-002-70493 Supplier Medchemexpress LLC Supplier No. HYW7225621MG
Trimethylamine oxide-15N is the deuterium labeled Trimethylamine N-oxide (HY-116084) Trimethylamine N-oxide is a gut microbe-dependent metabolite of dietary choline and other trimethylamine-containing nutrients Trimethylamine N-oxide induces inflammation by activating the ROS/NLRP3 inflammasome Trimethylamine N-oxide also accelerates fibroblast-myofibroblast differentiation and induces cardiac fibrosis by activating the TGF- /smad2 signaling pathway[1][2][3]