Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution

Catalog No. p-4510501
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC370785000 500 mL Glass bottle
AC370780010 1 L Glass bottle
AC370780025 2.5 L Glass bottle
AC370780100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC370785000 Supplier Thermo Scientific Chemicals Supplier No. 370785000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 75-50-3 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylamine
Color Colorless
CAS 75-50-3, 7732-18-5
Boiling Point 30.0°C to 100.0°C
Melting Point -2.0°C
Molecular Formula C3H9N
Linear Formula (CH3)3N
MDL Number MFCD00008327
Density 0.8600g/mL
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether, benzene, toluene,, xylene, ethylbenzene and chloroform
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
SMILES CN(C)C
IUPAC Name N,N-dimethylmethanamine
Molecular Weight (g/mol) 59.11
PubChem CID 1146
ChEBI CHEBI:18139
Formula Weight 59.11
Percent Purity 48 to 52% (total base)
Grade Pure
Packaging Glass bottle
Specific Gravity 0.86
Quantity 500 mL
Flash Point −45°C
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.