Learn More
Medchemexpress LLC Trimethylammonium chloride-13C3 | 286013-00-1 | 98.0% | 98.55 | 13C3H10ClN | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW5859055MG
Encompass_Preferred
Trimethylammonium chloride-13C3 is the carbon-13-labeled form of trimethylammonium chloride, supplied as a research-grade analytical standard for use as an internal standard, tracer, or reference in mass spectrometry and metabolic studies. The material is provided in a small, accurately measured quantity suitable for analytical workflows.
  • Isotopically labeled (13C3) for reliable mass spectrometry quantitation.
  • Suitable as a tracer in metabolic and analytical studies.
  • Supplied as a 5 mg vial for analytical applications.
  • Reported molecular weight 98.55 for calculation and method development.
  • High reported purity (98.0%) supporting analytical accuracy.
Catalog No. 50-002-84225