Learn More
Medchemexpress LLC Trimethylammonium chloride | 593-81-7 | MFCD00012478 | 95.57 g/mol | C3H10ClN | 50mg
SDP
Catalog No. 5000269074
Encompass_Preferred
Trimethylammonium chloride (Standard) is the analytical standard of Trimethylammonium chloride This product is intended for research and analytical applications Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]
Catalog No. 50-002-69074 Supplier Medchemexpress LLC Supplier No. HYY0504R50MG
Trimethylammonium chloride (Standard) is the analytical standard of Trimethylammonium chloride This product is intended for research and analytical applications Trimethylammonium chlorideIt is an endogenous metabolite that inhibits deacetylation Trimethylammonium chlorideIs a non-competitive inhibitor of acetylcholinesterase[1][2]