Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylboroxine

Catalog No. 903965250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-396-5250ML 5 g
90-396-525ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 90-396-5250ML Supplier Sigma Aldrich Supplier No. 903965250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylboroxine (TMB) is a cyclic anhydride of methyl-boronic acid. It can be used as a:Methylating agent for the methylation of various aromatic halides and C(sp3)-H bonds using palladium catalyst.Reagent in the preparation of polymer supported CBS (Corey, Bakshi, and Shibata) catalysts.Organoboranes. 39. Convenient procedures for the preparation of methylboronic acid and trimethylboroxin

Specifications

CAS 2362-50-7
Molecular Formula C12H8OS2
Refractive Index n/D 1.3880
Linear Formula C12H8OS2
MDL Number MFCD01464038
Quantity 5 g
Synonym Non-fluorinated sulfoxide-based thianthrene reagent; Thianthrene 5-oxide
Molecular Weight (g/mol) 232.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.