Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl 1,3,5-benzenetricarboxylate, 99%, Thermo Scientific Chemicals

Catalog Number AC152070250
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
2672-58-4
C12H12O6
252.22
MFCD00008434
RGCHNYAILFZUPL-UHFFFAOYSA-N
trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
75881
trimethyl benzene-1,3,5-tricarboxylate
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
2672-58-4
C12H12O6
252.22
MFCD00008434
RGCHNYAILFZUPL-UHFFFAOYSA-N
trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
75881
trimethyl benzene-1,3,5-tricarboxylate
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 144°C to 147°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(CO2CH3)3
Quantity 25 g
Beilstein 09, 979
Formula Weight 252.22
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Trimethyl 1,3,5-benzenetricarboxylate
Show More Show Less
EINECSNumber 220-215-5

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit