Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylene oxide

Catalog No. T762015G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T762015G 5 g
T7620110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T762015G Supplier Sigma Aldrich Supplier No. T762015G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trimethylene oxide is a strained cyclic ether which can be used to replace carbonyl / gem-dimethyl groups during molecular modelling of bioactive molecules
  • It can be incorporated in various bioactive molecule and drug substances for its improved physicochemical properties, via novel c-h activation methods
  • Trimethylene oxide can also be used in the synthesis of poly(trimethylene oxide) and related polymer electrolytes

Specifications

CAS 503-30-0
Density 0.893 g/mL (at 25°C (literature))
Boiling Point 50°C (lit.)
Molecular Formula C3H6O
Refractive Index n20/D 1.388-1.396; n20/D 1.392 (literature)
Linear Formula C3H6O
Quantity 5 g
MDL Number MFCD00005167
Synonym 1,3-Epoxypropane; 1,3-Propylene oxide; Cyclooxabutane; Oxacyclobutane; Oxetane
Molecular Weight (g/mol) 58.08
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.