Learn More
Strem, An Ascensus Company CAS# 1825-62-3. 100g. Trimethylethoxysilane, min. 97%. MFCD00009069
SDP
Catalog No. 5090114154
Encompass
CAS# 1825-62-3. 100g. Trimethylethoxysilane, min. 97%. MFCD00009069. Molecular Weight: 118.30. Molecular Formula: (CH3)3SiOC2H5. Color/form: colorless liq. Strem# 93-1465. http://www.strem.com/catalog/v/93-1465/
Catalog No. 50-901-14154 Supplier Strem, An Ascensus Company Supplier No. 931465100G
CAS# 1825-62-3. 100g. Trimethylethoxysilane, min. 97%. MFCD00009069. Molecular Weight: 118.30. Molecular Formula: (CH3)3SiOC2H5. Color/form: colorless liq. Strem# 93-1465. http://www.strem.com/catalog/v/93-1465/