Learn More
Strem, An Ascensus Company CAS# 1445-79-0. 25g. Trimethylgallium, 99+%. MFCD00014841
SDP
Supplier:  Strem, An Ascensus Company 31200025G
Encompass
CAS# 1445-79-0. 25g. Trimethylgallium, 99+%. MFCD00014841. Molecular Weight: 114.83. Molecular Formula: (CH3)3Ga. Color/form: colorless liq. Strem# 31-2000. http://www.strem.com/catalog/v/31-2000/
Catalog No. 50-204-5298