Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Trimethylhydroquinone, 98%
Click to view available options
Quantity:
500g
Description
Applications
Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.
Solubility
Soluble in water (2 g/L @ 20°C.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.
Solubility
Soluble in water (2 g/L @ 20°C.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications
Melting Point | 169°C to 174°C |
Quantity | 500g |
Flash Point | 140°C (284°F) |
UN Number | UN3077 |
Beilstein | 1909183 |
Solubility Information | Soluble in water (2g/L @ 20°C. |
Formula Weight | 152.2 |
Percent Purity | 98% |
Chemical Name or Material | Trimethylhydroquinone |
Safety and Handling
GHS H Statement
H318-H332-H317-H335-H315
Causes serious eye damage.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
P261-P262-P264b-P270-P271-P272-P280-P302+P350-P304+P340-P305+P351+P338-P310-P312-P333+P313-P361-P363-P501c
H310-H315-H317-H318-H332-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 211-838-3
RTECSNumber : MX7820000
TSCA : Yes