Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyloctadecylammonium Bromide

Catalog No. 35924610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-924-610G 10 g
35-924-650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-924-610G Supplier Sigma Aldrich Supplier No. 35924610G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethyloctadecylammonium Bromide

Specifications

CAS 1120-02-1
Melting Point ∼250°C
Molecular Formula C21H46BrN
Linear Formula CH3(CH2)17N(Br)(CH3)3
MDL Number MFCD00043171
Quantity 10 g
Synonym C18TAB; TMODAB; Octadecyltrimethylammonium bromide
Molecular Weight (g/mol) 392.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.