Learn More
Medchemexpress LLC Trimethyloctylammoni 25g
SDP
Catalog No. 5000205714
Encompass_Preferred
Trimethyloctylammonium bromide (TOAB) is used as a surfactant and phase transfer catalyst in various chemical reactions TOAB can be used in the synthesis of nanomaterials due to its ability to selectively transfer ions across interfaces and as a surfactant in the production of emulsions and foams It is valued for its amphiphilic properties which allow it to interact with water and oils stabilizing and dispersing mixtures
Catalog No. 50-002-05714 Supplier Medchemexpress LLC Supplier No. 25GHYW099642
Trimethyloctylammonium bromide (TOAB) is used as a surfactant and phase transfer catalyst in various chemical reactions TOAB can be used in the synthesis of nanomaterials due to its ability to selectively transfer ions across interfaces and as a surfactant in the production of emulsions and foams It is valued for its amphiphilic properties which allow it to interact with water and oils stabilizing and dispersing mixtures