Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyloctylammonium Chloride

Catalog No. 750945G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-094-5G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 75-094-5G Supplier Sigma Aldrich Supplier No. 750945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethyloctylammonium chloride (TOMAC) is a cationic surfactant that can be used: To separate SiC fro mground SiO2 by flotation method. To prepare porous organosilicates using 1,2-bis(trimethoxysilyl)ethane as a silica precursor.

Specifications

CAS 147409-41-4
Melting Point 185°C to 190°C
Molecular Formula C22H26N4O2
Linear Formula C22H26N4O2
MDL Number MFCD22417223
Quantity 1 g
Synonym Octyltrimethylammonium chloride
Molecular Weight (g/mol) 378.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.