Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylolpropane diallyl ether

Catalog No. 416126250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-612-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-612-6250ML Supplier Sigma Aldrich Supplier No. 416126250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylolpropane diallyl ether (TMPAE) is an unsaturated allyl ether that can be synthesized by Williamson esterification reaction in the presence of trimethylolpropane, sodium hydroxide, and allyl chloride as starting materials.TMPAE can be used in the synthesis of isophorone diisocyanate (IPDI) based tetracene monomers which can further be used in the formation of polymer dispersed liquid crystals. It can also be used in the synthesis of allyl ether based dendrimers that find potential usage in coatings.remainder monoethers and triethers

Specifications

CAS 682-09-7
Density 0.955 g/mL (at 25°C)
Boiling Point 135°C (13 mmHg)
Molecular Formula C12H22O3
Refractive Index n20/D 1.458 (literature)
Linear Formula C2H5C(CH2OCH2CH=CH2)2 CH2OH
Quantity 250 mL
MDL Number MFCD00035791
Synonym 2,2-Bis(allyloxymethyl)-1-butanol
Molecular Weight (g/mol) 214.3
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.