Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylolpropane ethoxylate

Catalog No. 4161771L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-617-71L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-617-71L Supplier Sigma Aldrich Supplier No. 4161771L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylolpropane ethoxylate

Specifications

CAS 50586-59-9
Density 1.1 g/mL (at 25°C)
Refractive Index n20/D 1.472
Linear Formula (C2H4O)n(C2H4O)n(C2H4O)nC6H14O
Quantity 1 L
MDL Number MFCD00197947
Synonym 3 arm PEG; 3 arm PEO; Poly(ethylene oxide), 3-arm, hydroxy terminated; Polyethylene glycol trimethylolpropane ether; Polyethylene glycol trimethylolpropane triether; Trimethylolpropane ethoxylate (20/3 EO/OH); Trimethylolpropane ethoxylate (4.6 EO/OH); Trimethylolpropane ethoxylate (4/15 EO/OH)
Molecular Weight (g/mol) 1014
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.