Learn More
Medchemexpress LLC Trimethylolpropane ethoxylate triacrylate | 28961-43-5 | 428.47 | 10 G
SDP
Catalog No. 5000351690
Encompass_Preferred
Trimethylolpropane ethoxylate triacrylate (ETPTA) is a plasticizer utilized in the synthesis of nanocomposites. It is intended for research use only.
  • Can be used for the fabrication of photonic crystals with different positions of the photonic stop band.
  • Achieves this by inverting opal-type templates using photopolymerization of ETPTA photocurable resin.
  • Allows gold nanoparticles to be embedded into inverse ETPTA films by multiple infiltration.
Catalog No. 50-003-51690 Supplier Medchemexpress LLC Supplier No. HY13944610G
Trimethylolpropane ethoxylate triacrylate (ETPTA) is a plasticizer utilized in the synthesis of nanocomposites. It is intended for research use only.
  • Can be used for the fabrication of photonic crystals with different positions of the photonic stop band.
  • Achieves this by inverting opal-type templates using photopolymerization of ETPTA photocurable resin.
  • Allows gold nanoparticles to be embedded into inverse ETPTA films by multiple infiltration.