Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylolpropane tris(3-mercaptopropionate)

Catalog No. 381489100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-148-9100ML 100 mL
38-148-9500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-148-9100ML Supplier Sigma Aldrich Supplier No. 381489100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trimethylolpropane tris(3-mercaptopropionate) is reported as a multi-thiol crosslinking reagent.trimethylolpropane tris(3-mercaptopropionate) may be used in the following studies:
  • Preparation of hydrogels, via crosslinking inulin derivatives
  • Construction of porous hybrid monolithic materials
  • As monomer for the preparation of a new thiol-ene based polymeric fluorescence sensor, via photo initiated polymerization

Specifications

CAS 33007-83-9
Density 1.21 g/mL (at 25°C (literature))
Boiling Point 220°C (0.3 mmHg)
Molecular Formula C15H26O6S3
Refractive Index n20/D 1.518 (literature)
Linear Formula (HSCH2CH2CO2CH2)3 CC2H5
MDL Number MFCD00046842
Quantity 100 mL
Molecular Weight (g/mol) 398.56
Percent Purity ≥95.0%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.