Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylolpropane tris[poly(propylene glycol), amine terminated] ether

Catalog No. 406708250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-670-8250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-670-8250ML Supplier Sigma Aldrich Supplier No. 406708250ML
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylolpropane tris[poly(propylene glycol), amine terminated] ether (T-403) is a type of polyetheramine that has a primary amine group. It can be used as an epoxy curing agent. T-403 has a hydroxyl group that reacts with the epoxy monomer to form stable ether linkages.T-403 may be surface functionalized by graphene nanoplatelet (GNP) and can be used as a water coolant in closed conduit flow. It can be incorporated into poly(butylene terephthalate) (PBT) as a blend, which enhances its melt-flowability. It can also be used as a cross-linker, which facilitates the curing of ethylene-glycidyl methacrylate copolymer.

Specifications

CAS 39423-51-3
Density 0.981 g/mL (at 25°C)
Linear Formula C2H5C[CH2[OCH2CH(CH3)]nNH2]3
Refractive Index n20/D 1.461 (literature)
MDL Number MFCD00197951
Quantity 250 mL
Molecular Weight (g/mol) 440
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.